Pectinase, Aspergillus niger
Pectinase, Aspergillus niger
Thermo Scientific Chemicals
Pectinase, Aspergillus niger
Used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number ALFJ63408.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS2517-43-3
IUPAC Name3-methoxybutan-1-ol
Molecular FormulaC5H12O2
InChI KeyJSGVZVOGOQILFM-UHFFFAOYNA-N
SMILESCOC(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Water=<0.2 %
Refractive index1.4150 to 1.4165 (20°C, 589 nm)
Appearance (Form)Liquid
View more
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.