Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Thermo Scientific Chemicals
Cytidine-5'-monophosphate disodium salt, 99+%
CAS: 6757-06-8 | C9H12N3Na2O8P | 367.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ALFJ63376.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS123-76-2
IUPAC Name4-oxopentanoic acid
Molecular FormulaC5H8O3
InChI KeyJOOXCMJARBKPKM-UHFFFAOYSA-N
SMILESCC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Refractive Index1.4395-1.4435 @ 20?C