Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Thermo Scientific Chemicals
Cytidine-5'-monophosphate disodium salt, 99+%
CAS: 6757-06-8 | C9H12N3Na2O8P | 367.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFJ63376.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7782-82-3
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Elemental Analysis(Selenium, Se) : 49.69-54.91% (Theory 52.31%)
Appearance (Color)White to pale cream to pale gray
Assay (unspecified)(By Selenium Elemental Analysis) : >95.0 to <105.0%