Ketoconazole, 98%, Thermo Scientific Chemicals
Ketoconazole, 98%, Thermo Scientific Chemicals
Ketoconazole, 98%, Thermo Scientific Chemicals
Ketoconazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ketoconazole, 98%, Thermo Scientific Chemicals
Potent inhibitor of cytochrome P450c17
Have Questions?
Catalog NumberQuantity
ALFJ63367.065 g
Catalog number ALFJ63367.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialKetoconazole
CAS65277-42-1
Health Hazard 1H301-H360F-H373
Health Hazard 2GHS H Statement
H301-H360-H373
Toxic if swallowed.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
View more
Ketoconazole is used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. Ketoconazole is an antifungal agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ketoconazole is used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. Ketoconazole is an antifungal agent.
Solubility
Soluble in DMSO, ethanol, chloroform, water, and methanol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Mikihisa Takano.; Risa Hasegawa.; Takeshi Fukuda.; Ryoko Yumoto.; Junya Nagai.; Teruo Murakami. Interaction with P-glycoprotein and transport of erythromycin, midazolam and ketoconazole in Caco-2 cells. European Journal of Pharmacology.1998, 358, (3), 289-294.
  2. Nick E Goeders.; Rachel L Peltier.; Glenn F Guerin. Ketoconazole reduces low dose cocaine self-administration in rats.Drug and Alcohol Dependence.1998, 53, (1), 67-77.