Phosphate, 0.5M buffer soln., pH 7.5
Phosphate, 0.5M buffer soln., pH 7.5
Phosphate, 0.5M buffer soln., pH 7.5
Phosphate, 0.5M buffer soln., pH 7.5
Thermo Scientific Chemicals
Phosphate, 0.5M buffer soln., pH 7.5
CAS: 7558-80-7119.976 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63349.AP500 mL
Catalog number ALFJ63349.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS40273-47-0
IUPAC Name3-fluoropyridine-4-carbaldehyde
Molecular FormulaC6H4FNO
InChI KeyKLKQXQXSZCNWLZ-UHFFFAOYSA-N
SMILESFC1=C(C=O)C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient Temperatures