Tricine-buffered saline (5X), pH 7.5, Thermo Scientific™
Tricine-buffered saline (5X), pH 7.5, Thermo Scientific™
Tricine-buffered saline (5X), pH 7.5, Thermo Scientific™
Tricine-buffered saline (5X), pH 7.5, Thermo Scientific™
Tricine-buffered saline (5X), pH 7.5, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFJ63294.AP500 mL
Catalog number ALFJ63294.AP
Price (MYR)
240.70
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
240.70
Add to cart
Specifications
Chemical Name or MaterialTricine-buffered saline
CAS5704-04-1
Recommended StorageAmbient temperatures
Concentration5X
TSCAYes
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.