BCA Protein Assay Kit
BCA Protein Assay Kit
Thermo Scientific Chemicals
BCA Protein Assay Kit
Have Questions?
Catalog NumberQuantity
ALFJ63283.QA1 Kit
Catalog number ALFJ63283.QA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Kit
Request bulk or custom format
Chemical Identifiers
CAS26385-07-9
IUPAC NameN-(2-chloroethyl)benzamide
Molecular FormulaC9H10ClNO
InChI KeyFYQJUYCGPLFWQR-UHFFFAOYSA-N
SMILESClCCNC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or fused solid
Assay (HPLC)≥96.0%
BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes. Accurately measures protein concentrations from 0.5 to 30 ?g/mL in tube assays and 1 to 20 ?g/mL in 96- or 384-well plate assays. It is used for studying protein: protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.