Piroxicam, Thermo Scientific Chemicals
Piroxicam, Thermo Scientific Chemicals
Piroxicam, Thermo Scientific Chemicals
Piroxicam, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piroxicam, Thermo Scientific Chemicals
CAS: 36322-90-4 | C15H13N3O4S | 331.346 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFJ63239.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1174064-57-3
IUPAC Name4-isocyanato-1-methyl-1H-pyrazole
Molecular FormulaC5H5N3O
InChI KeyTXMLNGHIFGKEFH-UHFFFAOYSA-N
SMILESCN1C=C(C=N1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain fine suspension
GC>=94 %
Appearance (Color)Yellow to orange
Appearance (Form)Liquid
Infrared spectrumConforms
Piroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.
Solubility
Soluble in water, ethanol, chloroform, ethyl acetate.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. V. Tantishaiyakul.; N. Kaewnopparat.; S. Ingkatawornwong. Properties of solid dispersions of piroxicam in polyvinylpyrrolidone K-30. International Journal of Pharmaceutics. 1996, 143,(1), 59-66.
  2. V. Tantishaiyakul.; N. Kaewnopparat.; S. Ingkatawornwong. Properties of solid dispersions of piroxicam in polyvinylpyrrolidone K-30. International Journal of Pharmaceutics. 1999, 181,(2), 143-151.