N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS, Thermo Scientific Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS, Thermo Scientific Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS, Thermo Scientific Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS, Thermo Scientific Chemicals
Used for colorimetric determination of sulfonamide in blood
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ALFJ63214.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS87-72-9
IUPAC Name(3S,4R,5R)-oxane-2,3,4,5-tetrol
Molecular FormulaC5H10O5
InChI KeySRBFZHDQGSBBOR-ZRMNMSDTSA-N
SMILESO[C@@H]1COC(O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=99.0 %
Water=<0.5 % (K.F.)
View more
Used for colorimetric determination of sulfonamide in bloodN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used for colorimetric determination of sulfonamide in bloodN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
Solubility
Soluble in water or 95% ethanol.Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.
Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
RUO – Research Use Only
General References:
  1. Refat, M. S.; Adam, A. M. A.; Sharshar, T.; Saad, H. A.; Eldaroti, H. H. Utility of positron annihilation lifetime technique for the assessment of spectroscopic data of some charge-transfer complexes derived from N-(1-Naphthyl)ethylenediamine dihydrochloride. Spectrochim. Acta, Part A 2014, 122, 34-47.
  2. Zhu, J.; Li, C.; Liu, S.; Liu, Z.; Yang, J.; Tian, J.; Hu, X. A non-diazotization-coupling reaction-based colorimetric determination of nitrite in tap water and milk. Eur. Food Res. Technol. 2014, 238 (5), 889-894.