Famotidine, 98+%, Thermo Scientific Chemicals
Famotidine, 98+%, Thermo Scientific Chemicals
Famotidine, 98+%, Thermo Scientific Chemicals
Famotidine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Famotidine, 98+%, Thermo Scientific Chemicals
A competitive histamine H2 receptor antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFJ63165.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS2353-45-9
IUPAC Namedisodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
Molecular FormulaC37H34N2Na2O10S3
InChI KeyXJBPDZVCYTYRIN-UHFFFAOYSA-L
SMILES[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
Assay (unspecified)Dye content: 85% min.
FormPowder
Solution Test10% in NaOH, blue solution
Famotidine, is commonly used in the treatment of peptic ulcer disease and gastroesophageal reflux disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water and methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. RP Walt.; MJS Langman.; J Cottrell.; SG Mann. Continuous intravenous famotidine for haemorrhage from peptic ulcer. The Lancet. 1992340 (8827), 1058-1062.
  2. M Jaimini.; AC Rana.; YS Tanwar. Formulation and evaluation of famotidine floating tablets. Current drug delivery. 20074 (1), 51-55.