Naproxen sodium, 98%, Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naproxen sodium, 98%, Thermo Scientific Chemicals
Naproxen sodium (CAS # 26159-34-2) is an arylacetic acid of the non-steroidal anti-inflammatory drug class.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFJ63103.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS536-17-4
IUPAC Name(5Z)-5-{[4-(dimethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular FormulaC12H12N2OS2
InChI KeyJJRVRELEASDUMY-YFHOEESVSA-N
SMILESCN(C)C1=CC=C(C=C2/SC(=S)NC2=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with TBAH98.5 to 101.5 %
Infrared spectrumConforms
Suitability(for Ag determination) Passes test
Appearance (Form)Powder
Solubility(0.03 % in acetone) Clear
View more
A cyclooxygenase inhibitorNaproxen sodium acts as a nonselective inhibitor of Cox-1 and Cox-2. It serves as an anti-inflammatory agent with analgesic and antipyretic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Naproxen sodium is the sodium salt of naproxen
  • Naproxen sodium competitively suppresses prostaglandin formation and pyrogen release by blocking the conversion of arachidonic acid
Application
  • It is suitable for cell culture research.
RUO – Research Use Only
General References:
  1. K Patwardhan, S.; J Patil, M.; Anbazhagan, S. Piper Betle: A Promising Tool for Drug Permeation Enhancement of Naproxen Sodium Transdermal Gel Formulation. Curr Drug ther 2015, 10 (1), 35-43.
  2. Ünlüer, Ö. B.; Genç, L.; Kahyaoğlu, Ş. G.; Ersöz, A. Controlled Release of Naproxen Sodium from Supermacroporous Cryogels. J. Pharm. Pharmacol. 2014, 2, 527-533.