Transfer or electro blotting buffer (10X), pH 8.5
Transfer or electro blotting buffer (10X), pH 8.5
Thermo Scientific Chemicals
Transfer or electro blotting buffer (10X), pH 8.5
Used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2L
1L
4L
Catalog number ALFJ63037.K3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2L
Request bulk or custom format
Chemical Identifiers
CAS1883-32-5
IUPAC Name2,2-diphenylethan-1-ol
Molecular FormulaC14H14O
InChI KeyNYLOEXLAXYHOHH-UHFFFAOYSA-N
SMILESOCC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
Melting point57°C to 61°C
Transfer or electro blotting buffer (10X), pH 8.5 is used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.