Transfer or electro blotting buffer (10X), pH 8.5
Transfer or electro blotting buffer (10X), pH 8.5
Thermo Scientific Chemicals
Transfer or electro blotting buffer (10X), pH 8.5
Used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1L
2L
4L
Catalog number ALFJ63037.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1L
Request bulk or custom format
Specifications
FormLiquid
Quantity1L
Storage RequirementsAmbient temperatures
Unit Size
Transfer or electro blotting buffer (10X), pH 8.5 is used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.