Antibiotic A23187, 98%
Antibiotic A23187, 98%
Antibiotic A23187, 98%
Antibiotic A23187, 98%
Thermo Scientific Chemicals
Antibiotic A23187, 98%
An antibiotic and calcium ionophore | CAS: 52665-69-7 | C29H37N3O6 | 523.63 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63020.MA10 mg
Catalog number ALFJ63020.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialAntibiotic A23187
TypeAntibiotic A23187
Physical FormPowder
CAS52665-69-7
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
A23187 (Mixed Calcium-Magnesium Salt) is used in cell activation experiments when calcium dose-response data are not required. Ca-Mg ratio ∼ 1:1. it is widely used as a compound in cell activation. It is used as a highly selective Ca2+ ionophore. it is used in studies of intracellular Ca2+ mediated events. A23187 has been observed to induce cell activation, differentiation or proliferation. In vitro experimentation has shown A23187 to boost ROS generation and platelet fragmentation. A23187 is a widely used ionophore. It has the ability to increase the intracellular concentrations of cations and is widely used to increase the intracellular levels of calcium. Visualization of Synaptic Ca2+ Calmodulin-Dependent Protein Kinase II Activity in Living Neurons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A23187 (Mixed Calcium-Magnesium Salt) is used in cell activation experiments when calcium dose-response data are not required. Ca-Mg ratio ∼ 1:1. it is widely used as a compound in cell activation. It is used as a highly selective Ca2+ ionophore. it is used in studies of intracellular Ca2+ mediated events. A23187 has been observed to induce cell activation, differentiation or proliferation. In vitro experimentation has shown A23187 to boost ROS generation and platelet fragmentation. A23187 is a widely used ionophore. It has the ability to increase the intracellular concentrations of cations and is widely used to increase the intracellular levels of calcium. Visualization of Synaptic Ca2+ Calmodulin-Dependent Protein Kinase II Activity in Living Neurons.
Solubility
Soluble in DMSO with warming or sonication. Also soluble in ethanol or ethyl acetate
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ding et al. Differential effects of endoplasmic reticulum stress-induced autophagy on cell survival. J.Biol.Chem.2007, 282, (7), 4702-10 .
  2. Kajitani et al. Mechanism of A23187-induced apoptosis in HL-60 cells: dependency on mitochondrial permeability transition but not NADPH oxidase. Biosci.Biotechnol.Bichem.2007, 22, (4), 888-93.