Anisomycin, 97%
Anisomycin, 97%
Anisomycin, 97%
Thermo Scientific Chemicals
Anisomycin, 97%
CAS: 22862-76-6 | C14H19NO4 | 265.31 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62964.MF50 mg
Catalog number ALFJ62964.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Chemical Identifiers
CAS621-23-8
IUPAC Name1,3,5-trimethoxybenzene
Molecular FormulaC9H12O3
InChI KeyLKUDPHPHKOZXCD-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=CC(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Appearance (Form)Powder or crystals or flakes and/or chunks
Infrared spectrumConforms
Melting point50°C to 54°C
GC>=98.5 %
A protein synthesis inhibitor. Causes apoptosis of PC12 Cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • A protein synthesis inhibitor
  • Causes apoptosis of PC12 Cells
RUO – Research Use Only