Ethylenediaminetetraacetic acid, Cell Culture Reagent
Ethylenediaminetetraacetic acid, Cell Culture Reagent
Ethylenediaminetetraacetic acid, Cell Culture Reagent
Ethylenediaminetetraacetic acid, Cell Culture Reagent
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, Cell Culture Reagent
CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number ALFJ62948.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.7-100.3% NaHCO₃ min
Impurity contentFe 0.001%
Impurity contentK 0.005%
Insoluble material0.015%
Heavy Metals(ICP-OES) 5ppm
View more
Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.
Solubility
Soluble in water.
Notes
Not tested for endtoxin or mycoplasma contaminants
RUO – Research Use Only
General References:
  1. Argandoña, M. et al. Interplay between iron homeostasis and the osmotic stress response in the halophilic bacterium Chromohalobacter salexigens. Appl. Environ. Microbiol.. 2010, 76(11), 3575-3589.
  2. Le Pichon, CE. et al. Expression and localization of the prion protein PrP(C) in the olfactory system of the mouse. J. Comp. Neurol. 2008, 508(3), 487-99.