Chromomycin A3, 97%
Chromomycin A3, 97%
Chromomycin A3, 97%
Thermo Scientific Chemicals
Chromomycin A3, 97%
CAS: 7059-24-7 | C57H82O26 | 1183.257 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62927.MA10 mg
Catalog number ALFJ62927.MA
Price (MYR)
2,745.00
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
2,745.00
Add to cart
Specifications
Chemical Name or MaterialChromomycin A3
TypeChromomycin A3
Physical FormPowder
CAS7059-24-7
Health Hazard 1H300-H360
View more
Chromomycin A3, is used as a G-C specific DNA ligand which inhibits transcription and acts as a DNA binding dye. Chromomycin A3 antagonizes enhanced DNA binding of the transcription factors Sp1 and Sp3 to their cognate G-C box, induced by oxidative stress or DNA damage. Furthermore, by inhibiting transcription, Chromomycin A3 and its structural analogs can inhibit protein biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chromomycin A3, is used as a G-C specific DNA ligand which inhibits transcription and acts as a DNA binding dye. Chromomycin A3 antagonizes enhanced DNA binding of the transcription factors Sp1 and Sp3 to their cognate ″G-C&Pr
RUO – Research Use Only
General References:
  1. CT Amemiya.;JR Gold. Chromomycin A3 stains nucleolus organizer regions of fish chromosomes. Copeia. 19861986 (1), 226-231.
  2. RH Jensen. Chromomycin A3 as a fluorescent probe for flow cytometry of human gynecologic samples. Journal of Histochemistry & Cytochemistry. 197725 (7), 573-579.