Aztreonam, Thermo Scientific Chemicals
Aztreonam, Thermo Scientific Chemicals
Aztreonam, Thermo Scientific Chemicals
Aztreonam, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aztreonam, Thermo Scientific Chemicals
CAS: 78110-38-0 | C13H17N5O8S2 | 435.426 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62887.031 g
Catalog number ALFJ62887.03
Price (MYR)
902.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
902.00
Add to cart
Specifications
Chemical Name or MaterialAztreonam
TypeAztreonam
Physical FormPowder
CAS78110-38-0
Melting Point227°C (decomposition)
View more
Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.
Solubility
Soluble in DMF/water (1:1) at 50 mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Dallal MM.; Czachor JS. Aztreonam-induced myelosuppression during treatment of Pseudomonas aeruginosa pneumonia. DICP. 1991, 25 (6), 594-7.
  2. Kobayashi, Y.; Uchida, H.; Kawakami Y. Synergy with aztreonam and arbekacin or tobramycin against Pseudomonas aeruginosa isolated from blood. J Antimicrob Chemother . 1992, 30 (6), 871-872.