Omeprazole, 98%, Thermo Scientific Chemicals
Omeprazole, 98%, Thermo Scientific Chemicals
Omeprazole, 98%, Thermo Scientific Chemicals
Omeprazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Omeprazole, 98%, Thermo Scientific Chemicals
A proton pump inhibitor
Have Questions?
Catalog NumberQuantity
ALFJ62860.065 g
Catalog number ALFJ62860.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS8005-03-6
IUPAC Namedisodium (3E)-5-amino-6-[(1E)-2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC22H14N6Na2O9S2
InChI KeyHKBVRFLHNUEVRO-HMDKGIQMSA-L
SMILES[Na+].[Na+].NC1=C2C(=O)\C(=N/NC3=CC=CC=C3)C(=CC2=CC(=C1\N=N\C1=CC=C(C=C1)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
lambda-max (UV-Vis)560-590nm (water)
Appearance (Color)Black
Specific Absorbance (E 1%/1cm)>200 at Lambda max (water)
Loss on Drying≤15% (105?C, constant weight)
View more
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.
Solubility
Soluble in water (0.5 mg/ml), DMSO (25 mg/ml), and ethanol (4.5 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Satoh et al. Antisecretory and anitulcer activities of a novel proton pump inhibitor AG-1749 in dogs and rats. J.Pharmacol.Exp.Ther. 1989, 248(2), 806-15.
  2. Schmarda et al. The gastric H,K-ATPase blocker lansoprazole is an inhibitor of chloride channels. Br.J.Pharmacol. 2000, 129(3), 598-604.