Trimyristin, 95%
Trimyristin, 95%
Trimyristin, 95%
Thermo Scientific Chemicals
Trimyristin, 95%
CAS: 555-45-3 | C45H86O6 | 723.177 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62819.1425 g
Catalog number ALFJ62819.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioning agent, solvents and viscosity controlling agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioni
RUO – Research Use Only
General References:
  1. A. van Langevelde, et al. Structure of [beta]-trimyristin and [beta]-tristearin from high-resolution X-ray powder diffraction data.Acta Cryst.,2001,B57372-377.
  2. Martins, S, et al. Improving Oral Absorption of Samon Calcitonin by Trimyristin Lipid Nanoparticles.Journal of Biomedical Nanotechnology.,2009,5(1), 76-83.