Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™
Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™
Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™
Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™
Thermo Scientific Chemicals
Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFJ62817.AE100 mL
Catalog number ALFJ62817.AE
Price (MYR)
385.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
385.00
Add to cart
Specifications
Chemical Name or MaterialPotassium acetate
ColorColorless
CAS127-08-2
Recommended StorageAmbient temperatures
TSCAYes
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.