NP-40 lysis buffer (2X), Thermo Scientific Chemicals
NP-40 lysis buffer (2X), Thermo Scientific Chemicals
Thermo Scientific Chemicals
NP-40 lysis buffer (2X), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ62805.AK250 mL
Catalog number ALFJ62805.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS580-20-1
IUPAC Namequinolin-1-ium-7-olate
Molecular FormulaC9H7NO
InChI KeyXCRPPAPDRUBKRJ-UHFFFAOYSA-N
SMILES[O-]C1=CC=C2C=CC=[NH+]C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point239°C to 244°C (Dec)
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient temperatures