View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAgarose gel loading dye
Name NoteFicoll based
Physical FormViscous Liquid
Concentration6X
Recommended StorageKeep cold