Agarose Gel Loading Dye (6X, Ficoll-Based)
Agarose Gel Loading Dye (6X, Ficoll-Based)
Agarose Gel Loading Dye (6X, Ficoll-Based)
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
25 mL
Catalog number ALFJ62800.AB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAgarose gel loading dye
Name NoteFicoll based
Physical FormViscous Liquid
Concentration6X
Recommended StorageKeep cold