(+/-)-Baclofen, 99+%
(+/-)-Baclofen, 99+%
(+/-)-Baclofen, 99+%
(+/-)-Baclofen, 99+%
Thermo Scientific Chemicals
(+/-)-Baclofen, 99+%
CAS: 1134-47-0 | C10H12ClNO2 | 213.661 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62768.031 g
Catalog number ALFJ62768.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-Baclofen
CAS1134-47-0
Health Hazard 1H301-H315-H317-H319-H334-H335-H360
Health Hazard 2GHS H Statement
H301-H334-H360-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
View more
Selective GABA-b receptor agonist