Mefenamic acid, 98%, Thermo Scientific Chemicals
Mefenamic acid, 98%, Thermo Scientific Chemicals
Mefenamic acid, 98%, Thermo Scientific Chemicals
Mefenamic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mefenamic acid, 98%, Thermo Scientific Chemicals
CAS: 61-68-7 | C15H15NO2 | 241.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFJ62705.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS68855-54-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
pH9.0 to 11.0 (10 % slurry)
Wet density255 to 370 g/l
Permeability2.6 to 6.5 Darcy
View more
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.
Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Hixson, L J., et al . Antiproliferative effect of nonsteroidal antiinflammatory drugs against human colon cancer cells. Cancer epidemiology, biomarkers & prevention . American Association for Cancer Research. 2002, 3, (5), 433-8.
  2. H Gögelein.; D Dahlem.; HC Englert.; HJ Lang. Flufenamic acid, mefenamic acid and niflumic acid inhibit single nonselective cation channels in the rat exocrine pancreas. FEBS letters. 1990, 268, (1), 79-82.