Phosphate-buffered saline (PBS, 10X), pH 7.6
Phosphate-buffered saline (PBS, 10X), pH 7.6
Thermo Scientific Chemicals
Phosphate-buffered saline (PBS, 10X), pH 7.6
Helps to maintain a constant pH
Have Questions?
Catalog NumberQuantity
ALFJ62692.K21 L
Catalog number ALFJ62692.K2
Price (MYR)
609.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
609.00
Add to cart
Specifications
Recommended StorageAmbient temperatures
Dilution10 X
Product TypePhosphate Buffered Saline (PBS)
Quantity1 L
pH7.6