TRIS-buffered saline (TBS, 10X) pH 7.6
TRIS-buffered saline (TBS, 10X) pH 7.6
TRIS-buffered saline (TBS, 10X) pH 7.6
TRIS-buffered saline (TBS, 10X) pH 7.6
Thermo Scientific Chemicals
TRIS-buffered saline (TBS, 10X) pH 7.6
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62662.K32 L
Catalog number ALFJ62662.K3
Price (MYR)
849.00
Add to cart
Quantity:
2 L
Request bulk or custom format
Price (MYR)
849.00
Add to cart
Specifications
Chemical Name or MaterialTRIS-buffered saline
CAS77-86-1
Recommended StorageAmbient temperatures
Concentration10X
TSCAYes
View more
TRIS-buffered saline (TBS, 10X) pH 7.6 is used as a substance diluent, as a wash buffer in immunoassays such as enzyme-linked immunosorbent assay. It is used in antibody diluent in western blotting, sample diluent in vitro diagnostics, immuno-histochemical staining and in situ hybridization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Miscible with water.
Notes
250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.6.Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhang, A.; Han, Y.; Wang, B.; Li, S.; Gan, W. Beyond Gap Junction Channel Function: the Expression of Cx43 Contributes to Aldosterone-Induced Mesangial Cell Proliferation via the ERK1/2 and PKC Pathways. Cell. Physiol. Biochem. 2015, 36 (3), 1210-1222.
  2. Shaoyuan, C.; Ming, D.; Yulang, H.; Hongcheng, F. Increased IL-37 in Atherosclerotic Disease could be Suppressed by Atorvastatin Therapy. Scand. J. Immunol. 2015, 82 (4), 328-336.