Trichloroacetic acid, 5% w/v aq. soln., Thermo Scientific Chemicals
Trichloroacetic acid, 5% w/v aq. soln., Thermo Scientific Chemicals
Trichloroacetic acid, 5% w/v aq. soln., Thermo Scientific Chemicals
Trichloroacetic acid, 5% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroacetic acid, 5% w/v aq. soln., Thermo Scientific Chemicals
Protein precipitation reagent
Have Questions?
Catalog NumberQuantity
ALFJ62366.AP500 mL
Catalog number ALFJ62366.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS629-03-8
IUPAC Name1,6-dibromohexane
Molecular FormulaC6H12Br2
InChI KeySGRHVVLXEBNBDV-UHFFFAOYSA-N
SMILESBrCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5056 to 1.5076 (20°C, 589 nm)
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.
Solubility
Soluble in water (1200 mg/ml at 20°C), alcohol, ether, ethanol (very soluble), and carbon tetrachloride.
Notes
Store under nitrogen. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases, smines.
RUO – Research Use Only
General References:
  1. RW Blakesley.; JA Boezi. A new staining technique for proteins in polyacrylamide gels using Coomassie Brilliant Blue G250. Analytical biochemistry. 1977, 82(2), 580-582.
  2. WH Marsh.; B Fingerhut.; H Miller. Automated and manual direct methods for the determination of blood urea. Clinical chemistry. 1965, 11(6), 624-627.