Trichloroacetic acid, 10% w/v aq. soln.
Trichloroacetic acid, 10% w/v aq. soln.
Trichloroacetic acid, 10% w/v aq. soln.
Trichloroacetic acid, 10% w/v aq. soln.
Thermo Scientific Chemicals
Trichloroacetic acid, 10% w/v aq. soln.
CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62355.AP500 mL
Catalog number ALFJ62355.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTrichloroacetic acid
CAS76-03-9
Health Hazard 1H314-H335-H351
ColorColorless
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, d
RUO – Research Use Only
General References:
  1. RW Blakesley.; JA Boezi. A new staining technique for proteins in polyacrylamide gels using Coomassie Brilliant Blue G250. Analytical biochemistry. 1977, 82(2), 580-582.
  2. WH Marsh.; B Fingerhut.; H Miller. Automated and manual direct methods for the determination of blood urea. Clinical chemistry. 1965, 11(6), 624-627.