beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals
CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFJ62337.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Materialbeta-Nicotinamide adenine dinucleotide
CAS53-84-9
ColorWhite
Recommended StorageStore at -20°C
RTECS NumberUU3450000
View more
NAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
NAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.
Solubility
Soluble in water at 50mg/ml
Notes
b-NAD is very hygroscopic and should be stored desiccated. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C.
RUO – Research Use Only
General References:
  1. Pollak N.; Dölle C.; Ziegler M. The power to reduce: pyridine nucleotides - small molecules with a multitude of functions. Biochem. J. 2007, 402(2), 205-18.
  2. Unden G, Bongaerts J. Alternative respiratory pathways of Escherichia coli: energetics and transcriptional regulation in response to electron acceptors. Biochim. Biophys. Acta. 1997, 1320(3), 217-34.