Ethidium bromide soln., 10mg/ml
Ethidium bromide soln., 10mg/ml
Ethidium bromide soln., 10mg/ml
Ethidium bromide soln., 10mg/ml
Thermo Scientific Chemicals
Ethidium bromide soln., 10mg/ml
CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62282.AB10 mL
Catalog number ALFJ62282.AB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Request bulk or custom format
Chemical Identifiers
CAS617-89-0
IUPAC Name1-(furan-2-yl)methanamine
Molecular FormulaC5H7NO
InChI KeyDDRPCXLAQZKBJP-UHFFFAOYSA-N
SMILESNCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4890 to 1.4910 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow-brown
GC>=99.0 %
Appearance (Form)Liquid
View more
Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.1,2,3 It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA.4 Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase. Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.