5(6)-Tetramethylrhodamine isothiocyanate
5(6)-Tetramethylrhodamine isothiocyanate
Thermo Scientific Chemicals
5(6)-Tetramethylrhodamine isothiocyanate
CAS: 95197-95-8 | C25H21N3O3S
Have Questions?
Catalog NumberQuantity
ALFJ62258.MA10 mg
Catalog number ALFJ62258.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS93-97-0
IUPAC Namebenzoyl benzoate
Molecular FormulaC14H10O3
InChI KeyCHIHQLCVLOXUJW-UHFFFAOYSA-N
SMILESO=C(OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
HPLC>=97.5 %
Melting point38°C to 45°C
Appearance (Color)White to cream to light beige
5(6)-TRITC is widely used in preparing bioconjugates of proteins and nucleic acids. It is suitable for protein, peptide and nucleotide labeling. Serves as an acceptor for FRET. It is recommended as a donor molecule in these FRET-based applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5(6)-TRITC is widely used in preparing bioconjugates of proteins and nucleic acids. It is suitable for protein, peptide and nucleotide labeling. Serves as an acceptor for FRET. It is recommended as a donor molecule in these FRET-bas
RUO – Research Use Only
General References:
  1. J.H. Kim, et al. Polymeric nanoparticles for protein kinase activity. Chem. Commun. 2007, 7 (13), 1346-8.
  2. M. Heidecker, et al. Proximity relationships and structural dynamics of the phalloidin binding site of actin filaments in solution and on single actin filaments on heavy meromyosin. Biochemistry. 1995, 34 (35), 11017-25.