Adenosine-3',5'-cyclic monophosphate sodium salt, 99%
Adenosine-3',5'-cyclic monophosphate sodium salt, 99%
Adenosine-3',5'-cyclic monophosphate sodium salt, 99%
Adenosine-3',5'-cyclic monophosphate sodium salt, 99%
Thermo Scientific Chemicals
Adenosine-3',5'-cyclic monophosphate sodium salt, 99%
CAS: 37839-81-9 | C10H11N5NaO6P | 351.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ALFJ62174.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialAdenosine-3',5'-cyclic monophosphate sodium salt
CAS37839-81-9
ColorWhite
Recommended StorageStore at -20°C
TSCANo
View more
Adenosine 3?,5?-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.
Solubility
Soluble in water (50 mg/ml).
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. W Epstein.; LB Rothman-Denes. Adenosine 3': 5'-cyclic monophosphate as mediator of catabolite repression in Escherichia coli. PNAS.1975, 72, (6), 2300-2304.
  2. CM Szego.; JS Davis. Adenosine 3', 5'-monophosphate in rat uterus: acute elevation by estrogen. PNAS.1967, 58, (4), 1711-1718.