Agarose Gel Loading Dye (6X, Alkaline)
Agarose Gel Loading Dye (6X, Alkaline)
Thermo Scientific Chemicals
Agarose Gel Loading Dye (6X, Alkaline)
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
25 mL
Catalog number ALFJ62157.AB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Request bulk or custom format
Chemical Identifiers
CAS12003-13-3
IUPAC Nametitanium(4+) aluminium(3+) heptachloride
Molecular FormulaAlCl7Ti
InChI KeyZMKAVICCBWPNSR-UHFFFAOYSA-G
SMILES[Al+3].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)TiCl₃ 76.0-78.5%
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
Solubility
Fully miscible in water.
Notes
300mM sodium hydroxide, 6mM EDTA, 0.25% bromocresol green, 0.25% xylene cyanol FF and 18% Ficoll
RUO – Research Use Only
General References:
  1. Georges F. Carle and Maynard V. Olson. Separation of chromosomal DNA molecules from yeast by orthogonal-field-alternation gel electrophoresis. Nucleic Acids Research.1984, 12 (14), 5647-5664.
  2. Robert P. Noble. Electrophoretic separation of plasma lipoproteins in agarose gel. The Journal of Lipid Research.1968, 9 693-700.