Sodium pyrophosphate, 200mM buffer soln.
Sodium pyrophosphate, 200mM buffer soln.
Sodium pyrophosphate, 200mM buffer soln.
Sodium pyrophosphate, 200mM buffer soln.
Thermo Scientific Chemicals
Sodium pyrophosphate, 200mM buffer soln.
CAS: 7758-16-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number ALFJ62052.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS3392-97-0
IUPAC Name2,6-dimethoxybenzaldehyde
Molecular FormulaC9H10O3
InChI KeyWXSGQHKHUYTJNB-UHFFFAOYSA-N
SMILESCOC1=CC=CC(OC)=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point95°C to 98°C
GC>=98.5 %
Appearance (Color)Yellow to beige
Appearance (Form)Crystalline powder
Sodium pyrophosphate, 200mM buffer soln. is used in the laboratory as a buffering agent. DSPP has been used as a chelating agent in antimicrobial studies. Tetrabasic pyrophosphate or TSPP (sc-203404; sc-236921) has shown to be useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. It is a popular leavening agent found in baking powders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium pyrophosphate, 200mM buffer soln. is used in the laboratory as a buffering agent. DSPP has been used as a chelating agent in antimicrobial studies. Tetrabasic pyrophosphate or TSPP (sc-203404; sc-236921) has shown to be useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. It is a popular leavening agent found in baking powders.
Solubility
Fully miscible in water. Insoluble in alcohol and ammonia.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Wei Zhao; Hui Yin; Fan Liu; Xiong-Han Feng; Wen-Feng Tan. [Characterization of Pb2+ adsorption on the surface of birnessite treatment with Na4P2O7 at different pH and the study on the distribution of Mn(III) in the birnessite]. Environmental Science.2011, 32, (8), 2477-2484.
  2. Sanko Nguyen; Marianne Hiorth; Morten Rykke; Gro Smistad. The potential of liposomes as dental drug delivery systems. European Journal of Pharmaceutics and Biopharmaceutics.2011, 77, (1), 75-83.
Contents & Storage
Ambient Temperatures