Ethidium bromide soln., 0.625mg/ml
Ethidium bromide soln., 0.625mg/ml
Ethidium bromide soln., 0.625mg/ml
Ethidium bromide soln., 0.625mg/ml
Thermo Scientific Chemicals
Ethidium bromide soln., 0.625mg/ml
CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62029.AA5 mL
Catalog number ALFJ62029.AA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialEthidium bromide soln.
ColorRed
CAS1239-45-8
Health Hazard 1H331-H341
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
View more
For visualization of DNA in gel electrophoresis