Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
CAS: 63-92-3 | C18H23Cl2NO | 340.288 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61900.MD250 mg
Catalog number ALFJ61900.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS101366-51-2
IUPAC Name4'-methyl-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular FormulaC13H11ClO2S
InChI KeyDBYXACYHGMSPMQ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98%
Selective alpha adrenergic blocking agent. Irreversible calmodulin inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Selective alpha adrenergic blocking agent
  • Irreversible calmodulin inhibitor
RUO – Research Use Only