Sodium chloride, 5M aq. soln., autoclaved
Sodium chloride, 5M aq. soln., autoclaved
Sodium chloride, 5M aq. soln., autoclaved
Sodium chloride, 5M aq. soln., autoclaved
Thermo Scientific Chemicals
Sodium chloride, 5M aq. soln., autoclaved
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61890.AK250 mL
Catalog number ALFJ61890.AK
Price (MYR)
447.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
447.00
Add to cart
Specifications
Chemical Name or MaterialSodium chloride
Name Noteautoclaved
CAS7647-14-5