Papain, Carica Papaya Latex
Papain, Carica Papaya Latex
Papain, Carica Papaya Latex
Papain, Carica Papaya Latex
Thermo Scientific Chemicals
Papain, Carica Papaya Latex
A cysteine protease; breaks down intercellular matrix of cartilage
Have Questions?
Catalog NumberQuantity
ALFJ61875.MB25 mg
Catalog number ALFJ61875.MB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Request bulk or custom format
Chemical Identifiers
CAS7681-65-4
IUPAC Nameλ¹-copper(1+) iodide
Molecular FormulaCuI
InChI KeyLSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES[Cu+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C