D-Lactic acid, Thermo Scientific Chemicals
D-Lactic acid, Thermo Scientific Chemicals
D-Lactic acid, Thermo Scientific Chemicals
D-Lactic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D-Lactic acid, Thermo Scientific Chemicals
Useful chiral synthon, building block for depsipeptides
Have Questions?
Catalog NumberQuantity
ALFJ61871.MD250 mg
Catalog number ALFJ61871.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS10378-47-9
IUPAC Nameλ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Molecular FormulaCeH20N4O18S4
InChI KeyVCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILESN.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Yellow to orange
Titration with Na2S2O3>=99.0 %
Iron (Fe)=<50 ppm
Insoluble matter=<0.02 % (in dilute H2SO4)
View more
D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.
Solubility
Soluble in water (50mg/ml)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only
General References:
  1. Kobayashi D.; Nozawa T.; Imai K.; Nezu J.; Tsuji A.; Tamai I. Involvement of human organic anion transporting polypeptide OATP-B (SLC21A9) in pH-dependent transport across intestinal apical membrane. J Pharmacol Exp Ther. 2003, 306, (2), 703-8.
  2. Nozawa T.; Imai K.; Nezu J.; Tsuji A.; Tamai I. Functional characterization of pH-sensitive organic anion transporting polypeptide OATP-B in human. J Pharmacol Exp Ther. 2003, 308, (2), 438-45.