Minoxidil, Thermo Scientific Chemicals
Minoxidil, Thermo Scientific Chemicals
Minoxidil, Thermo Scientific Chemicals
Minoxidil, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Minoxidil, Thermo Scientific Chemicals
Minoxidil, CAS # 38304-91-5, is a comopund with vasodilating effects that works by opening adenosine triphosphate (ATP)-sensitive potassium channels.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFJ61803.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point132°C to 135°C
Appearance (Form)Crystals
Infrared spectrumConforms
Titration with HClO499.0 to 100.5 %
Heavy metals (as Pb)=<10 ppm
View more
Minoxidil is used as antihypertensive, antialopecia agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
  • Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure
RUO – Research Use Only
General References:
  1. Proctor PH. Endothelium-derived relaxing factor and minoxidil: active mechanisms in hair growth. Arch Dermatol. 1989 Aug;125(8):1146. PMID: 2757417.
  2. Nagai N, Iwai Y, Sakamoto A, Otake H, Oaku Y, Abe A, Nagahama T. Drug Delivery System Based On Minoxidil Nanoparticles Promotes Hair Growth In C57BL/6 Mice. Int J Nanomedicine. 2019 Oct 1;14:7921-7931. doi: 10.2147/IJN.S225496. PMID: 31632009; PMCID: PMC6778480.