Haloperidol
Haloperidol
Haloperidol
Haloperidol
Thermo Scientific Chemicals
Haloperidol
CAS: 52-86-8 | C21H23ClFNO2 | 375.87 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61688.065 g
Catalog number ALFJ61688.06
Price (MYR)
788.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
788.00
Add to cart
Specifications
Chemical Name or MaterialHaloperidol
CAS52-86-8
Health Hazard 1H301-H315-H317-H319-H335-H360
Health Hazard 2GHS H Statement
H301-H360-H315-H319-H317-H335
Toxic if swallowed.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
View more
An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats.
RUO – Research Use Only