Calciferol, Thermo Scientific Chemicals
Calciferol, Thermo Scientific Chemicals
Calciferol, Thermo Scientific Chemicals
Calciferol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calciferol, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ61610.031 g
Catalog number ALFJ61610.03
Price (MYR)
350.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
350.00
Add to cart
Specifications
Chemical Name or MaterialCalciferol
CAS50-14-6
Health Hazard 1H301+H311-H330-H372
Health Hazard 2GHS H Statement
H300-H310-H372
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
View more
Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.
Solubility
Soluble in ethanol, water, methanol, dimethylformamide, and dimethyl sulfoxide.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Gui-Dong. Zhu.; William H. Okamura. Synthesis of Vitamin D (Calciferol). Chem. Rev. 1995, 95, (6), 1877-1952.
  2. Maryka H. Bhattacharyya.; H.F. DeLuca. Subcellular location of rat liver calciferol-25-hydroxylase. Archives of Biochemistry and Biophysics. 1974, 160, (1), 58-62.