Clozapine, 97%, Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Potent, selective muscarinic antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Catalog number ALFJ61583.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS111-67-1
IUPAC Name(2Z)-oct-2-ene
Molecular FormulaC8H16
InChI KeyILPBINAXDRFYPL-HYXAFXHYSA-N
SMILESCCCCCC=C/C
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Assay (GC)> 97.5% (sum of cis and trans isomers)
Refractive Index1.4130 - 1.4170 @20?C
Appearance (Color)Clear, colourless
Potent, selective muscarinic antagonist