Clozapine, 97%, Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Clozapine, 97%, Thermo Scientific Chemicals
Potent, selective muscarinic antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number ALFJ61583.MB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialClozapine
CAS5786-21-0
Health Hazard 1H301-H315-H319-H335-H341-H361
Health Hazard 2GHS H Statement
H301-H361-H315-H319-H335
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
Potent, selective muscarinic antagonist