TRIS, 1.0M buffer soln., pH 7.8
TRIS, 1.0M buffer soln., pH 7.8
TRIS, 1.0M buffer soln., pH 7.8
TRIS, 1.0M buffer soln., pH 7.8
Thermo Scientific Chemicals
TRIS, 1.0M buffer soln., pH 7.8
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61579.K21 L
Catalog number ALFJ61579.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialTRIS buffer soln.
CAS77-86-1
Recommended StorageAmbient temperatures
Concentration1.0 M
TSCAYes