Bradford Dye Reagent, Ready-to-Use soln.
Bradford Dye Reagent, Ready-to-Use soln.
Thermo Scientific Chemicals
Bradford Dye Reagent, Ready-to-Use soln.
Suitable for protein concentration determination (100 to 1500μg)
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALFJ61522.K2
Price (MYR)
1,176.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,176.00
Add to cart
Specifications
Chemical Name or MaterialBradford Dye Reagent
ColorBlue
Name NoteReady-to-Use soln.
Health Hazard 1H290-H302+H332-H314-H335-H371
Health Hazard 2GHS H Statement
H314-H318-H371
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs.
View more
The Bradford Reagent is suitable for protein concentration determination (100-1500μg).Bradford dye reagent is used in the determination of protein content in cell fractions and assessing the protein concentrations.