Neomycin sulfate hydrate, Thermo Scientific Chemicals
Neomycin sulfate hydrate, Thermo Scientific Chemicals
Neomycin sulfate hydrate, Thermo Scientific Chemicals
Neomycin sulfate hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Neomycin sulfate hydrate, Thermo Scientific Chemicals
CAS: 1405-10-3 | C23H48N6O17S | 712.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ61499.22
Price (MYR)
942.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
942.00
Add to cart
Specifications
Chemical Name or MaterialNeomycin sulfate hydrate
TypeNeomycin Sulfate
Physical FormPowder
CAS1405-10-3
Health Hazard 1H317-H334-H361d-H373
View more
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.
Solubility
Soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only
General References:
  1. Nandi Zhou.; Jingyuan Wang.; Juan Zhang.; Can Li.; Yaping Tian.; Joseph Wang. Selection and identification of streptomycin-specific single-stranded DNA aptamers and the application in the detection of streptomycin in honey. Talanta. 2013, 108 109-116.
  2. Xiaojing Bai.; Hui Hou.; Bailin Zhang.; Jilin Tang. Label-free detection of kanamycin using aptamer-based cantilever array sensor. Biosensors and Bioelectronics. 2014, 56, 112-116.