Eserine, Thermo Scientific™
Eserine, Thermo Scientific™
Eserine, Thermo Scientific™
Eserine, Thermo Scientific™
Eserine, Thermo Scientific™
Inhibitor of acetylcholinesterase that crosses the blood-brain barrier
Have Questions?
Catalog NumberQuantity
ALFJ61477.MC100 mg
Catalog number ALFJ61477.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialEserine
Melting Point102°C to 104°C
CAS57-47-6
Health Hazard 1H300+H330
Health Hazard 2GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Eserine is a parasympathomimetic, specifically, a reversible cholinesterase inhibitor obtained from the Calabar bean, used to treat glaucoma and delayed gastric emptying. This compound has been used as a positive control in a study
General References:
  1. Nguyen PV.; Aniksztejn L.; Catarsi S.; Drapeau P. Maturation of neuromuscular transmission during early development in zebrafish. J Neurophysiol. 1999, 81, (6), 2852-61.
  2. Tuovinen K.; Kaliste-Korhonen E.; Raushel FM.; Hanninen O. Success of pyridostigmine, physostigmine, eptastigmine and phosphotriesterase treatments in acute sarin intoxication. Toxicology. 1999, 134, (2-3), 169-78.